Imprimer
Cryogenic Tape for Frozen Containers
Fournisseur: GA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TFNG19SBC150WH TFNG19SBC150BL TFNG19SBC150OR TFNG19SBC150GA TFNG19SBC150RE TFNG19SBC150YE TFNG25SBC150WH TFNG25SBC150BL TFNG25SBC150OR TFNG25SBC150GA TFNG25SBC150RE TFNG25SBC150YE
GAINTFNG19SBC150WHEA 36.2 EUR
GAINTFNG19SBC150WH GAINTFNG19SBC150BL GAINTFNG19SBC150OR GAINTFNG19SBC150GA GAINTFNG19SBC150RE GAINTFNG19SBC150YE GAINTFNG25SBC150WH GAINTFNG25SBC150BL GAINTFNG25SBC150OR GAINTFNG25SBC150GA GAINTFNG25SBC150RE GAINTFNG25SBC150YE
Cryogenic Tape for Frozen Containers
Étiquettes
Cryogenic tape for already frozen surfaces with GatorCUT™ technology. Can be cut by hand, without the need for any cutting instruments and applied directly on various frozen containers (no thawing required). Perfect for labeling containers for long-term preservation in liquid nitrogen and lab freezers.
  • Temperature range: −196 to 100 °C (−321 to 212 °F)
  • Ultra-quick application, conveniently portable so it can be easily carried for use whenever and wherever needed
  • Can be applied directly on various frozen containers (no thawing required), without the need of an overlap
  • Easy-tear GatorCUT™ technology; no more need for cutting instruments (dispensers, scissors, etc.)
  • Excellent water resistance; will retain adhesion even after exposure to condensation during thawing and immersion in water baths
This cryogenic tape is especially designed to be applied directly, and rapidly adhere, to frozen cryo vials, tubes, polycarbonate cryo boxes, plastic and cardboard freezer boxes, and other cryogenic containers which are difficult to label under deep-freeze conditions (−80 °C/−112 °F). Featuring GA internationals patented GatorCUT™ technology to effortlessly hand tear as little or as much needed without needing a tape dispenser or scissors. Designed for long-term preservation in liquid nitrogen (−196 °C/−321 °F), −80 °C (−112 °F) freezers, and dry ice transportation, they can also withstand water exposure such as thawing in water baths, as well as temperatures up to 100 °C (212 °F). Can be easily written on with permanent cryo-markers.