Imprimer
Nuclear and cytoplasmic extraction reagents, NE-PER
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
78833 78835
78833.EA 391 EUR
78833. PIER78835
Nuclear and cytoplasmic extraction reagents, NE-PER
Tampons Tampons d'extraction
The NE-PER Nuclear and Cytoplasmic Extraction Kit provides for efficient cell lysis and extraction of separate cytoplasmic and nuclear protein fractions in less than two hours. The nuclear protein extraction method involves simple, stepwise lysis of cells and centrifugal isolation of nuclear and cytoplasmic protein fractions. A benchtop microcentrifuge, tubes and pipettors are the only tools required. The NE-PER Reagents efficiently solubilise and separate cytoplasmic and nuclear proteins into fractions with minimal cross-contamination or interference from genomic DNA and mRNA. The isolated proteins can be used to perform immunoassays as well as protein interaction studies such as mobility shift assays (EMSA), co-immunoprecipitation (Co-IP) and pull-down assays.
  • Fast - obtain nuclear and cytoplasmic fractions in less than two hours
  • Proven - the NE-PER Reagent Kit is referenced in more than 950 distinct publications
  • Scalable - two kit sizes for producing extracts from cells and tissues
  • Convenient - simple instructions do not require ultracentrifugation over gradients
  • Compatible - use for downstream assays, including Western blotting, gel-shift assays, protein assays, reporter gene assays and enzyme activity assays
Informations de livraison: Kit contains Cytoplasmic Extraction Reagent I (CER I), 10 ml; Cytoplasmic Extraction Reagent II (CER II), 0.55 ml; Nuclear Extraction Reagent (NER), 5 ml.