Imprimer
Objectives EC Epiplan
Fournisseur: ZEISS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4220309902000 4220409961000 4220509961000 4220709961000 4220909961000
ZEIS4220309902000EA 0 EUR
ZEIS4220309902000 ZEIS4220409961000 ZEIS4220509961000 ZEIS4220709961000 ZEIS4220909961000
Objectives EC Epiplan
Objectifs pour microscopes
Enhanced contrast universal objectives for materials microscopy allowing brightfield, differential interference contrast and polarisation methods. HD versions provide darkfield capability in addition. They feature the long working distances needed for materials inspections.
  • Thread type: M27×0,75
  • Immersion: Without immersion
  • Field of view: 23 mm
  • Parfocal length: 45 mm
Optical system: Infinity Color Corrected System (ICS)
Color correction: Achromatic