Imprimer
Memosens Oxygen Sensors for the Food and Life Sciences Industries, COS22E
Fournisseur: Endress+Hauser
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
COS22E-19Q8/0 COS22E-1369/0
ENDRCOS22E-19Q8/0EA 3840 EUR
ENDRCOS22E-19Q8/0 ENDRCOS22E-1369/0
Memosens Oxygen Sensors for the Food and Life Sciences Industries, COS22E
Électrodes Sondes à oxygène dissous
Hygienic, amperometric oxygen sensor for the food and life sciences industries.
  • Accurate measurements with long-term stability and permanent self-monitoring
  • Application-specific versions available
  • The inductive Memosens coupling is insensitive to moisture and corrosion
The Memosens oxygen sensor COS22E is fully compliant to FDA and USP and thus suitable for hygienic and sterile applications. The sensor is low-maintenance and ensures long-term stability - even with frequent sterilization. Its trace version safeguards your products, process and plant equipment from oxygen influences. Thanks to Memosens 2.0 digital technology, COS22E offers extended storage of calibration and process data providing the perfect basis for predictive maintenance and efficient operation.
Material: FKM O-ring, stainless steel cap