Imprimer
ESD Safety Sandals, Joran Low S1PL
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
710801/36 710801/37 710801/38 710801/39 710801/40 710801/41 710801/42 710801/43 710801/44 710801/45 710801/46 710801/47 710801/48
ELTN710801/36EA 93.9 EUR
ELTN710801/36 ELTN710801/37 ELTN710801/38 ELTN710801/39 ELTN710801/40 ELTN710801/41 ELTN710801/42 ELTN710801/43 ELTN710801/44 ELTN710801/45 ELTN710801/46 ELTN710801/47 ELTN710801/48
ESD Safety Sandals, Joran Low S1PL
Chaussures et bottes
ESD safety shoe made from microfibre/textile upper and a rubber/TPU/EVA sole (Transformers).
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full-length inlay sole ESD
  • Non-metallic, penetration-resistant midsole
  • Rubber scuff cap
  • Composite toe cap
Areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Black/red
Certifications: EN ISO 20345 S1PL FO/HRO/SC/SR, form A.