Imprimer
ESD Safety Ankle Boots, Lacing Sytem, Stefano XXSG Pro Boa® GTX Mid S3
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
768751/38 768751/39 768751/40 768751/41 768751/48 768751/36 768751/37 768751/42 768751/43 768751/44 768751/45 768751/46 768751/47
ELTN768751/38EA 145.9 EUR
ELTN768751/38 ELTN768751/39 ELTN768751/40 ELTN768751/41 ELTN768751/48 ELTN768751/36 ELTN768751/37 ELTN768751/42 ELTN768751/43 ELTN768751/44 ELTN768751/45 ELTN768751/46 ELTN768751/47
ESD Safety Ankle Boots, Lacing Sytem, Stefano XXSG Pro Boa® GTX Mid S3
Chaussures et bottes
ESD safety ankle boot made from cowhide leather upper and a PU/PU sole (Wellmaxx Safety Grip).
  • All weather Gore-Tex performance membrane
  • Full, padded bellows tongue
  • Full-length inlay sole ARTISAN ESD
  • Boa® fit lacing system
  • Non-metallic, penetration-resistant midsole
  • PU toe protection
  • Steel toe cap
Suitable for areas where exposure to moisture is expected (S2). Areas where there is a risk of penetration from pointed and sharp objects (S3). Areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Black/blue
Certifications: EN ISO 20345 S3 CI/SRC, form B.