Imprimer
ESD Safety Ankle Boots, Lacing Sytem, Terence XXG Pro Pro Boa® Mid S3
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
768891/39 768891/40 768891/41 768891/42 768891/43 768891/44 768891/48 768891/45 768891/46 768891/47
ELTN768891/39EA 149.9 EUR
ELTN768891/39 ELTN768891/40 ELTN768891/41 ELTN768891/42 ELTN768891/43 ELTN768891/44 ELTN768891/48 ELTN768891/45 ELTN768891/46 ELTN768891/47
ESD Safety Ankle Boots, Lacing Sytem, Terence XXG Pro Pro Boa® Mid S3
Chaussures et bottes
ESD safety ankle boot made from cowhide leather uper and a rubber/PU sole (Wellmaxx Grip).
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • Boa® fit lacing system
  • Non-metallic, penetration-resistant midsole
  • Composite toe cap
  • TPU scuff cap
Indoors and outdoors. Areas where exposure to moisture is expected (S2). Areas where there is a risk of penetration from pointed and sharp objects (S3). Areas where there is a risk of electrostatic discharge (ESDS/ESD). Workplaces on hard undergrounds: The revolutionary Infinergy® sole core cushions impacts and provides for a rebound effect when the compressive impulse subsides - for more energy in every step.
Colour: Black
Certifications: EN ISO 20345 S3S WR/FO/SC/HI/HRO/M/SR, form B.