Imprimer
ESD Occupational Ankle Boots, Lace-Up, Enya XXF GTX Mid O2 WR CI
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
974670/35 974670/36 974670/37 974670/38 974670/39 974670/40 974670/41 974670/42
ELTN974670/35EA 124.9 EUR
ELTN974670/35 ELTN974670/36 ELTN974670/37 ELTN974670/38 ELTN974670/39 ELTN974670/40 ELTN974670/41 ELTN974670/42
ESD Occupational Ankle Boots, Lace-Up, Enya XXF GTX Mid O2 WR CI
Chaussures et bottes
ESD work boot made from hydrophobised suede and hydrophobised textile upper and a PU/PU sole (Wellmaxx Flex).
  • Full, padded bellows tongue
  • All weather Gore-Tex performance membrane
  • Full-length inlay sole ESD PRO Lady grey
  • Soft fleece ESD insole
Suitable for indoors and outdoors, areas where exposure to moisture is expected (O2), areas where there is no risk of falling heavy objects, areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Grey
Certifications: EN ISO 20347 O2 WR/FO/CI/SR, form B.