Imprimer
ESD Safety Shoes, Lacing System, Terence XXG Pro Boa® M Low S3 S HI
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
728891/45 728891/39 728891/40 728891/41 728891/42 728891/43 728891/44 728891/46 728891/47 728891/48
ELTN728891/45EA 0 EUR
ELTN728891/45 ELTN728891/39 ELTN728891/40 ELTN728891/41 ELTN728891/42 ELTN728891/43 ELTN728891/44 ELTN728891/46 ELTN728891/47 ELTN728891/48
ESD Safety Shoes, Lacing System, Terence XXG Pro Boa® M Low S3 S HI
Chaussures et bottes
ESD safety shoe made from cowhide uppper and a rubber/PU sole (Wellmaxx Grip).
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • Boa® fit lacing system
  • Non-metallic, penetration-resistant midsole
  • Composite toe cap
  • TPU scuff cap
Areas where there is a risk of electrostatic discharge (ESDS/ESD). Workplaces on hard undergrounds: The revolutionary Infinergy® sole core cushions impacts and provides for a rebound effect when the compressive impulse subsides - for more energy in every step.
Colour: Black
Certifications: EN ISO 20345 S3S WR/FO/SC/HRO/HI/M/SR, form A.