Imprimer
ESD Safety Shoes, Lace-Up, Ian XXTP Pro Low S3 S
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
721321/45 721321/46 721321/47 721321/48 721321/49 721321/35 721321/36 721321/37 721321/38 721321/39 721321/40 721321/41 721321/42 721321/43 721321/44
ELTN721321/45EA 99.9 EUR
ELTN721321/45 ELTN721321/46 ELTN721321/47 ELTN721321/48 ELTN721321/49 ELTN721321/35 ELTN721321/36 ELTN721321/37 ELTN721321/38 ELTN721321/39 ELTN721321/40 ELTN721321/41 ELTN721321/42 ELTN721321/43 ELTN721321/44
ESD Safety Shoes, Lace-Up, Ian XXTP Pro Low S3 S
Chaussures et bottes
ESD safety shoe made from hydrophobised microfibre/hydrophobised textile upper and a TPU/PU sole (Wellmaxx Trainers Power).
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • Non-metallic, penetration-resistant midsole
  • Composite toe cap
  • TPU scuff cap
Suitable for areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Grey
Certifications: EN ISO 20345 S3S FO/SR/SC, form A.