Imprimer
ESD Safety Shoes, Lacing Sytem, Ian XXTP Pro Boa® Low S3 S
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
721331/35 721331/36 721331/37 721331/40 721331/38 721331/39 721331/41 721331/42 721331/43 721331/44 721331/45 721331/46 721331/47 721331/48 721331/49
ELTN721331/35EA 118.9 EUR
ELTN721331/35 ELTN721331/36 ELTN721331/37 ELTN721331/40 ELTN721331/38 ELTN721331/39 ELTN721331/41 ELTN721331/42 ELTN721331/43 ELTN721331/44 ELTN721331/45 ELTN721331/46 ELTN721331/47 ELTN721331/48 ELTN721331/49
ESD Safety Shoes, Lacing Sytem, Ian XXTP Pro Boa® Low S3 S
Chaussures et bottes
ESD safety shoe is made from hydrophobised microfibre/hydrophobised textile upper and a TPU/PU sole (Wellmaxx Trainers Power). Sole core made of Infinergy® by BASF
  • Breathable fabric lining and heel pocket lining
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • Boa® fit lacing system
  • Non-metallic, penetration-resistant midsole
  • Composite toe cap
  • TPU scuff cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). The shoes comply to the standard 61340-5-1.
Colour: Blue
Certifications: EN ISO 20345:2022 S3S FO/SR/SC, form A.