Imprimer

Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (PerCP)

Fournisseur: AAT BIOQUEST
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

101611T0 101611T1 101611T2
AATB101611T0EA 271 EUR
AATB101611T0 AATB101611T1 AATB101611T2
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (PerCP)
Anticorps
The 3G8 monoclonal antibody recognises human CD16a, a 50 to 65 kD transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Attention: For research use only.

Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR