Imprimer
Manual Spray Dispensers
Fournisseur: Rubbermaid Commercial Products
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
RSZ5607 RSZ5107
RUBBRSZ5607EA 53.6 EUR
RUBBRSZ5607 RUBBRSZ5107
Manual Spray Dispensers
Distributeurs de savon et distributeurs de savon liquide
Best value safety in smaller spaces. Efficient dosing technology gives up to 4000 hand washes per 800 ml refill.
  • Simple spray solution produces a thick lather for thorough hand coverage
  • Hygienically sealed refills keep soap and hand sanitisers fresh and avoid cross contamination, and are fast and easy to service
  • Efficient dosing technology gives up to 4000 hand washes per refill
  • Antimicrobial pushbar is protected with BioMaster technology
  • Durable dispensers come with a lifetime warranty
Available refills include an assortment of convenient refill spray pouch and box designs, which are easy to install.
Choose from:
Alcohol Plus Hand Sanitiser: effective and fast acting, kills 99,99% of common germs.
Spray Lotion Soap with Moisturisers: rich, gentle hand wash with light fragrances for everyday use.