Imprimer
AutoCollect™ Specimen Collection Cards
Fournisseur: Ahlstrom-Munksjö
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8.560.0003.B-N 8.295.0003.B-N 8.297.0004.F-N 8.560.0006.B-N 8.298.0006.F-N 8.296.0003.B-N 8.561.0004.B-N
BINZ8.560.0003.B-NEA 451 EUR
BINZ8.560.0003.B-N BINZ8.295.0003.B-N BINZ8.297.0004.F-N BINZ8.560.0006.B-N BINZ8.298.0006.F-N BINZ8.296.0003.B-N BINZ8.561.0004.B-N
AutoCollect™ Specimen Collection Cards
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons biologiques
Ahlstrom AutoCollect™ specimen collection cards for high-throughput environments work efficiently in both DBS punch and direct elution. They are designed for microvolume sampling, transport and storage of dried blood spot (DBS) at ambient temperature.
  • Pharmacokinetic
  • Therapeutic drugs monitoring
  • Preclinical and clinical studies
  • Human identification
  • Toxicology
  • Forensics
  • Biomarkers screening
  • Viral load testing
The cassette format of the card with high stiffness provides a suitable fit with main laboratory automated equipment available on the market (including for example Camag DBS-MS 500, Hamilton EasyPunch and Spark Holland DBS Autosampler), allowing reliable DBS card processing, from detection of dried blood spot to the preparation of samples ready for LC/MS analysis.
Sample areas/card: 4
Packaging: 100 cards/pack
Informations de livraison: Supplied with instructions for use.
Cdt: The cards are shrinked-wrapped and packaged in a cardboard box.