Imprimer

Anti-BI 836845 Recombinant Antibody

Fournisseur: ProSci
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PRSI10-747EA 627 EUR
PRSI10-747
Anti-BI 836845 Recombinant Antibody
Anticorps
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG4-kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR