Imprimer
Réactif d'extraction de protéines, BugBuster® (sans amine primaire), Millipore®
Marques : Merck Millipore (Novagen)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
70923-3 70923-4
70923-3EA 179 EUR
70923-3 70923-4
Réactif d'extraction de protéines, BugBuster® (sans amine primaire), Millipore®
Tampons Tampons d'extraction
BugBuster® (primary amine-free) Protein Extraction Reagent is a special formulation of BugBuster designed for applications where primary amines would interfere if present in the protein extract, such as protein immobilisation or cross-linking. The PIPPS buffer used in BugBuster® (primary amine-free) has a similar buffer capacity and pH range as the original Tris-buffered BugBuster®, but will not complex metal ions, also making it ideally suited for extraction of metal dependent proteins.
Storage: 15 to 30 °C