Imprimer

Anti-Chicken IgY Goat Polyclonal Antibody (CF®583R)

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

20898-500UL 20898-50UL
20898-500ULEA 248 EUR
20898-500UL 20898-50UL
Anti-Chicken IgY Goat Polyclonal Antibody (CF®583R)
Anticorps
This is a highly cross-adsorbed goat anti-chicken IgY (H + L) secondary antibody labeled with our bright and photostable CF® dye. To minimize cross-reactivity, the antibody has been adsorbed against bovine, goat, guinea pig, Syrian hamster, horse, human, mouse, rabbit, rat, and sheep serum.

  • Highly cross-adsorbed for specific staining with minimal background
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues

Type: Secondary
Antigen: Chicken IgY
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Goat
Isotype: IgY
Reactivity: Chicken
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR