Imprimer
perform® classic concentrate Mucasol, liquid concentrate, universal detergent
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
44103 44105
148-0169EA 516 EUR
148-0169 148-0170
perform® classic concentrate Mucasol, liquid concentrate, universal detergent
Produits nettoyants Détergents et Nettoyants de Surfaces
Liquid, alkaline high-performance concentrate for immersion and ultrasonic baths. Universal cleaner for laboratory equipment and instruments made of glass, porcelain, plastic, rubber and metal.
  • Items are cleaned in ~ 10 – 30 minutes, without mechanical help
perform® classic concentrate mucasol replaces chromosulfuric acid, is free from chlorine and caustic alkalis, is non-toxic, nonaggressive and biodegradable. Prolonged exposure of zinc and non-anodized aluminium surfaces should be avoided