Minoxidil sulphate ≥98%
Marques : ENZO LIFE SCIENCESTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Attention
|
K⁺ channel activator.
A selective ATP-sensitive K⁺ (KATP) channel opener. It is the active metabolite of minoxidil and is a potent (IC₅₀=0.14 µM) vascular smooth muscle relaxant. Antihypertensive. Stimulates renin secretion.
Soluble in DMSO (25 mg/ml).
Formule:
C₉H₁₅N₅O₄S Température de stockage: Température ambiante |
Numéro MDL:
MFCD06248907 Numéro CAS: 83701-22-8 |
Résultats des tests de spécification
Purity | ≥98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...