Imprimer
Rapamycin 98 HPLC_ASSAY_METHOD
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): (3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-Hexadecahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,27-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentone
BMLA2750005 BMLA2750025
ENZOBMLA2750005EA 165 EUR
ENZOBMLA2750005 ENZOBMLA2750025
Rapamycin 98 HPLC_ASSAY_METHOD
Rapamycin
Macrocyclic-triene antibiotic possessing potent immunosuppressant activity. It forms a complex with FKBP12 that binds to an effector, thus inhibiting IL-2 and other growth promoting lymphokines. The effectors were identified as FRAP (FKBP12 rapamycin-associated protein) and RAFT1 (rapamycin and KFBP12 target). Rapamycin/FKBP complex does not inhibit FRAP PI 4-kinase activity, but does inhibit FRAP autophosphorylation. Rapamycin induces inhibition of p70s6k, p33cdk2 and p34cdc2. Selectively blocks signaling leading to the activation of p70/85 S6 kinase. Enhances apoptosis. Activator of autophagy both in vitro and in vivo.
  • Off-white or yellow solid
Soluble in chloroform, DMSO (25 mg/mL) or methanol
Cdt: Shipping temperature: Ambient
Formule: C₅₁H₇₉NO₁₃
Poids moléculaire: 914.19 g/mol
Température de stockage: Congélateur
Numéro MDL: MFCD00867594
Numéro CAS: 53123-88-9