Imprimer
Deoxynivalenol Fusarium sp. 98 HPLC, TLC
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): 3α,7α,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, DON, Vomitoxin
ALX630115M001 ALX630115M005
ENZOALX630115M001EA 93.5 EUR
ENZOALX630115M001 ENZOALX630115M005
Deoxynivalenol Fusarium sp. 98 HPLC, TLC
Deoxynivalenol
Mycotoxin found in cereals. Binds to the ribosome and inhibits protein synthesis.
Emetic and refusal activity in pig.
  • Mycotoxin
White to off-white powder.
Soluble in methanol or ethyl acetate; slightly soluble in water.
Formule: C₁₅H₂₀O₆
Poids moléculaire: 296.3 g/mol
Température de stockage: Réfrigérateur
Numéro CAS: 51481-10-8