Imprimer

8-(4-Chlorophenylthio)guanosine-3',5'-cyclic monophosphate sodium salt ≥99% (par HPLC)

Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ALX480088M001 ALX480088M005
ENZOALX480088M001EA 49.5 EUR
ENZOALX480088M001 ENZOALX480088M005
8-(4-Chlorophenylthio)guanosine-3',5'-cyclic monophosphate sodium salt ≥99% (par HPLC)
8-(4-Chlorophenylthio)guanosine-3',5'-cyclic monophosphate sodium salt

Selective activator of cGMP-dependent protein kinase (PKG) Iα, Iβ, and type II as well as of cGMP-gated ion channels.


Higher activation potential as compared to cGMP; excellent cell membrane permeability and phosphodiesterase stability. Preferred alternative to unsuitable analogs such as dibutyryl-cGMP.


Soluble in water.

Formule: C₁₆H₁₄ClN₅O₇PS·Na
Poids moléculaire: 509.8 g/mol
Température de stockage: Congélateur
Numéro CAS: 51239-26-0

Commander maintenant

Résultats des tests de spécification

Purity (HPLC) ≥99%
Identity Determined by ESI-MS and UV/VIS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR