Imprimer
8-Bromoadenosine 3',5'-cyclic Monophosphothioate, Rp-Isomer sodium salt ≥99% (par HPLC)
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ALX480024M001 ALX480024MC05
ENZOALX480024M001EA 382 EUR
ENZOALX480024M001 ENZOALX480024MC05
8-Bromoadenosine 3',5'-cyclic Monophosphothioate, Rp-Isomer sodium salt ≥99% (par HPLC)
8-Bromoadenosine 3',5'-cyclic Monophosphothioate, Rp-Isomer sodium salt
Inhibitor of cAMP-dependent protein kinase (PKA), preferentially binding to type I PKA. Lipophilic analog of Rp-cAMPS. Resistant to mammalian cyclic nucleotide-dependent phosphodiesterases. Significantly more lipophilic and membrane permeant as compared to Rp-cAMPS or 8-Br-cAMP.
  • PKA inhibitor
White to off-white powder.
250 mM soluble in water, aqueous solutions, DMSO or dimethyl formamide.
Formule: C₁₀H₁₀BrN₅O₅PS·Na
Poids moléculaire: 423.2 · 23.0 g/mol
Température de stockage: Congélateur
Numéro CAS: 129735-00-8