Imprimer
DMNQ ≥99% (par CCM)
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
ALX420027M005 ALX420027M010 ALX420027M025
ENZOALX420027M005EA 125 EUR
ENZOALX420027M005 ENZOALX420027M010 ENZOALX420027M025
DMNQ ≥99% (par CCM)
DMNQ
Inducer of intracellular ROS generation.
Redox-cycling agent that induces intracellular superoxide anion formation and, depending on the concentration, induces cell proliferation, apoptosis or necrosis. DMNQ does not react with free thiol groups, is non-alkylating and adduct-forming in contrast to other quinones. Thus, DMNQ is a valuable tool for the generation of reactive oxygen species (ROS) in order to study the role of ROS in cell toxicity, apoptosis and necrosis.
Soluble in acetone, DMSO or methanol.
Formule: C₁₂H₁₀O₄
Température de stockage: Congélateur
Numéro MDL: MFCD00184331
Numéro CAS: 6956-96-3
Commander maintenant
Résultats des tests de spécification
Purity ≥99% (TLC)
Identity Determined by NMR