Imprimer
Reveromycin A (de Streptomyces sp. MST-AS5344) ≥99% (par HPLC)
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ALX380216MC01 ALX380216MC25
ENZOALX380216MC01EA 271 EUR
ENZOALX380216MC01 ENZOALX380216MC25
Reveromycin A (de Streptomyces sp. MST-AS5344) ≥99% (par HPLC)
Reveromycin A
Antibiotic. Dominant analog of the reveromycin complex. Inhibitor of the mitogenic activity of epidermal growth factor (EGF). G1 phase cell cycle inhibitor, selectively inhibiting isoleucyl-tRNA synthetase. Displays antiproliferative as well as potent antifungal activity. Induces apoptosis in osteoclasts thus inhibiting bone resorption.
White to tan solid
Soluble in ethyl acetate, 100% ethanol, methanol, dimethyl formamide or DMSO
Cdt: Shipping temperature: Ambient
Formule: C₃₆H₅₂O₁₁
Température de stockage: Congélateur
Numéro CAS: 134615-37-5