Imprimer
Streptozocine 97
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): 1-Methyl-1-nitroso-3-((2S,3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)urea, Streptozotocin
ALX3800105100 ALX380010G001 ALX380010M100
ENZOALX3800105100EA 136 EUR
ENZOALX3800105100 ENZOALX380010G001 ENZOALX380010M100
Streptozocine 97
Streptozocine
Widely used diabetes inducer in rodents. Inhibition of β-cell O-GlcNAcase by streptozotocin is the mechanism that accounts for its diabetogenic toxicity. N-nitroso-containing antibiotic, acting as a nitric oxide (NO) donor and vasorelaxant. Relaxes phenylephrine-contracted aortic rings at 10µM. Potent methylating agent for DNA.
  • O-GlcNAcase inhibitor
White to pale yellow crystalline solid.
Soluble in water (nH2O at 102.8mg/ml) or 100% ethanol (200 proof at 0.92mg/ml).
Formule: C₈H₁₅N₃O₇
Poids moléculaire: 265.2 g/mol
Température de stockage: Congélateur
Numéro MDL: MFCD00006607
Numéro CAS: 18883-66-4
EINECS: 242-646-8