Imprimer
SB 225002 ≥98% (par HPLC)
Marques : Enzo Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ALX270478M001 ALX270478M005
ENZOALX270478M001EA 60.5 EUR
ENZOALX270478M001 ENZOALX270478M005
SB 225002 ≥98% (par HPLC)
SB 225002
Chemokine receptor ligand
Potent and selective antagonist of CXCR2 chemokine receptor (IC50=22nM). >150-fold more selective over CXCR1 receptor. Inhibits IL-8- (IC50=8nM) and GROα-mediated (IC50=10nM) calcium mobilisation. Prevents IL-8-induced neutrophil migration. Inhibits HIV replication in lymphocytes and macrophages.
Formule: C₁₃H₁₀BrN₃O₄
Numéro MDL: MFCD00954637
Numéro CAS: 182498-32-4