Imprimer
PD 184,352 ≥95% (par HPLC, RMN)
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): 2-(2-Chloro-4-iodoanilino)-N-(cyclopropylmethoxy)-3,4-difluorobenzamide, 2-((2-Chloro-4-iodophenyl)amino)-N-(cyclopropylmethoxy)-3,4-difluorobenzamide, PD 184352, CI-1040
ALX270471M001 ALX270471M005
ENZOALX270471M001EA 149 EUR
ENZOALX270471M001 ENZOALX270471M005
PD 184,352 ≥95% (par HPLC, RMN)
PD 184,352
Potent and selective inhibitor of MAPK (ERK kinase 1; MEK1) activation (IC50=300nM in vitro, IC50=2nM in vivo). Suppresses activation of MAPK but does not block its activity. Strongly suppresses growth of human implanted colon tumors in mice.
Synthetic.
Formule: C₁₇H₁₄ClF₂IN₂O₂
Numéro MDL: MFCD02683961
Numéro CAS: 212631-79-3