Imprimer
Pifithrin-α (cyclic) hydrobromide ≥95% (par HPLC)
Marques : Enzo Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ALX270287M010 ALX270287M050
ENZOALX270287M010EA 242 EUR
ENZOALX270287M010 ENZOALX270287M050
Pifithrin-α (cyclic) hydrobromide ≥95% (par HPLC)
Pifithrin-α (cyclic) hydrobromide
p53 inhibitor
Stable analog of pifithrin-alpha (Prod. No. BML-GR325), with biological properties similar to its parent compound, but with reduced cytotoxicity. Inhibitor of p53. Reversibly inhibits p53-dependent transactivation of p53-responsive genes and reversibly blocks p53-mediated apoptosis. Potent STAT6 transcriptional inhibitor.
Numéro CAS: 60477-34-1