Imprimer
DL-α-Difluoromethylornithine hydrochloride monohydrate ≥98% (par CCM)
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ALX270283M010 ALX270283M050
ENZOALX270283M010EA 52.8 EUR
ENZOALX270283M010 ENZOALX270283M050
DL-α-Difluoromethylornithine hydrochloride monohydrate ≥98% (par CCM)
DL-α-Difluoromethylornithine hydrochloride monohydrate
Inhibitor of ornithine decarboxylase and arginase
Specific, irreversible inhibitor of ornithine decarboxylase (ODC), the rate-limiting enzyme in polyamine biosynthesis. Potent chemopreventive agent. Induces apoptosis. Arginase activity inhibitor.
Numéro CAS: 68278-23-9