Imprimer
Captopril ≥98%
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): (S)-(-)-1-(3-Mercapto-2-methyl-1-oxopropyl)-L-proline, (S)-(-)-1-(3-Mercapto-2-methylpropionyl)-L-proline, (S)-1-((S)-3-mercapto-2-methylpropanoyl)pyrrolidine-2-carboxylic acid
ALX270212G001 ALX270212G005
ENZOALX270212G001EA 49.5 EUR
ENZOALX270212G001 ENZOALX270212G005
Captopril ≥98%
Captopril
Inhibitor of ACE and leukotriene hydrolase
First orally active angiotensin-converting enzyme (ACE) inhibitor. IC50=2.14nM. Reversible and competitive inhibitor of LTA4 hydrolase (IC50=11µM). Inhibits angiogenesis. Antihypotensive agent.
Formule: C₉H₁₅NO₃S
Numéro MDL: MFCD00168073
Numéro CAS: 62571-86-2