Imprimer
α-Toluenesulfonyle fluorure ≥99% (par GC-FID)
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): PMSF, Fluorure de toluène-α-sulfonyle
ALX270184G005 ALX270184G025
ENZOALX270184G005EA 94.6 EUR
ENZOALX270184G005 ENZOALX270184G025
α-Toluenesulfonyle fluorure ≥99% (par GC-FID)
α-Toluenesulfonyle fluorure
Irreversible inhibitor of serine proteases. Widely used fatty acid amid hydrolase (FAAH) inhibitor for pretreatment of brain membrane preparations when testing CB1 receptor activity. Spleen membranes used for CB2 studies do not require this pretreatment, as FAAH activity is absent in these cells. Inhibits also cysteine proteases like papain (reversible by DTT treatment, as well as internucleosomal DNA fragmentation in immature thymocytes. For a related, more stable inhibitor see AEBSF.
  • Inhibitor of serine proteases and FAAH
White to off-white needle crystals.
Soluble in 100% ethanol, methanol or isopropanol; insoluble in water.
Formule: C₇H₇FO₂S
Point d'ébullition: 80…85 °C (0,3 mmHg)
Point de fusion: 91…92 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00007424
Numéro CAS: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8,III