Imprimer

Arachidonoyl-AMC ≥99%

Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BMLST5040005 BMLST5040025
ENZOBMLST5040005EA 114 EUR
ENZOBMLST5040005 ENZOBMLST5040025
Arachidonoyl-AMC ≥99%
Arachidonoyl-AMC

FAAH substrate.


Fluorogenic substrate for fatty acid amide hydrolase (FAAH). This substrate is the basis for a simple, selective and sensitive assay for FAAH which is amenable to high-throughput screening.


Soluble in DMSO (>25 mg/ml).

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea8145f99e0dcf1eeb6f64901a4fe55210a72c0b&vid=1eecb728e3f8b07aa6fda7543ad36db9d80970e7&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Purity ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR