Imprimer
D,L-threo-PDMP hydrochloride ≥98% (par CCM)
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): D,L-threo-PDMP hydrochloride
BMLSL2100010 BMLSL2100050
ENZOBMLSL2100010EA 67.1 EUR
ENZOBMLSL2100010 ENZOBMLSL2100050
D,L-threo-PDMP hydrochloride ≥98% (par CCM)
D,L-threo-PDMP hydrochloride
Blocks glucosylation of ceramide by inhibiting UDP-glucose:ceramide glucosyltransferase (glucosylceramide synthetase). Inhibits the expression of cell surface glycolipid antigens and inhibits the growth of cultured rabbit skin fibroblasts. Possesses antitumor activity and inhibits Lewis lung carcinoma cell metastasis in vitro. Useful tool for studying the effects of cellular glycosphingolipid depletion. Enhances the apoptotic response to ionizing radiation by enhancing the ceramide signal.
  • Glucosylceramide synthetase inhibitor
White to off-white solid.
25mg/ml soluble in 100% ethanol, DMSO or water (50°C, vortex or sonicate).
Formule: C₂₃H₃₈N₂O₃·HCl
Température de stockage: Congélateur
Numéro MDL: MFCD00236490
Numéro CAS: 80938-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=feeb4ff60f78e4225484e7229b76e9b32250a07d&vid=938ee1e14b4cff6c06da680ddd364e4ec244d413&prid=web&clid=SAPDAM