Imprimer
Furoxan ≥99% (par HPLC)
Marques : Enzo Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BMLCN2550010 BMLCN2550050
ENZOBMLCN2550010EA 86.9 EUR
ENZOBMLCN2550010 ENZOBMLCN2550050
Furoxan ≥99% (par HPLC)
Furoxan
A novel NO donor which only releases NO in the presence of thiols. It is a potent vasodilator and inhibitor of platelet aggregation.
  • Releases NO in the presence of thiols
White solid.
Soluble in ethanol (25 mg/ml) or DMSO (25 mg/ml).
Formule: C₉H₅N₃O₂
Point de fusion: 72…74 °C
Température de stockage: Congélateur
Numéro CAS: 125520-62-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05e55818bd31a92ea2de1fc15e47ddad2165dd1d&vid=ab97e42541b4aa85e67daa78f9d929992923e1b1&prid=web&clid=SAPDAM