Imprimer
Prolyl endopeptidase inhibitor ≥97% (par HPLC)
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BMLPI1120005 BMLPI1120025
ENZOBMLPI1120005EA 91.3 EUR
ENZOBMLPI1120005 ENZOBMLPI1120025
Prolyl endopeptidase inhibitor ≥97% (par HPLC)
Prolyl endopeptidase inhibitor
A potent and selective inhibitor of prolyl endopeptidase (PEP/PREP/POP, EC 3.4.21.26). Ki=0.35 nM for mouse brain enzyme. Inhibits the cleavage of arginine-vasopressin, gonadotropin-releasing hormone, and other peptides.
  • Endopeptidase inhibitor
White solid.
Soluble in DMSO.
Formule: C₁₈H₂₂N₂O₄
Température de stockage: Congélateur
Numéro MDL: MFCD00890691
Numéro CAS: 86925-97-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3523aaf52707c6bbe1c89b26bb15107772f21da3&vid=f9c6933b7ccf566d540cd9fe6d5131179ba799b0&prid=web&clid=SAPDAM