Imprimer
Decylubiquinone ≥98% (par CCM)
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BMLCM1150010 BMLCM1150050
ENZOBMLCM1150010EA 90.2 EUR
ENZOBMLCM1150010 ENZOBMLCM1150050
Decylubiquinone ≥98% (par CCM)
Decylubiquinone
MPTP inhibitor
Inhibits the Ca2+ dependent mitochondrial permeability transition pore (MPTP) without inhibiting respiration. Maximum effect on Ca2+ retention capacity of rat liver mitochondria at 100μM. Inhibits etoposide-induced apoptosis in in L929 cells at 5μM.
Numéro CAS: 55486-00-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=766a9476c7a8d3f22efafa43ff50fe68eac75b2d&vid=086ba947302ecac492332d221e3d041dc09127e0&prid=web&clid=SAPDAM