Imprimer
IM-12 ≥98% (par HPLC)
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): 3-((4-Fluorophenethyl)amino)-1-methyl-4-(2-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione
BMLWN1020005 BMLWN1020025
ENZOBMLWN1020005EA 114 EUR
ENZOBMLWN1020005 ENZOBMLWN1020025
IM-12 ≥98% (par HPLC)
IM-12
GSK-3β inhibitor (IC50=53nM (enzymatic), 3.8µM (cellular)). The compound significantly increases β-catenin levels, leads to TCF-induction and promotes neuronal differentiation in human neural progenitor cells via activation of canonical Wnt signaling.
Formule: C₂₂H₂₀FN₃O₂
Poids moléculaire: 377.4115 g/mol
Numéro MDL: MFCD20527313
Numéro CAS: 1129669-05-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=379f66907c07eb41e3f5cfa40421ff00f4afd0c0&vid=e2ccfffa4984214f9220537943b9ac54d79beed6&prid=web&clid=SAPDAM