Imipramine-N-oxide ≥98% (par CCM), Ultrapure
Marques : ENZO LIFE SCIENCESTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
Imipramine metabolite produced via flavin-containing monooxygenase-mediated oxidation.
Soluble in DMSO (30 mg/ml), ethanol (20 mg/ml) or water (2 mg/ml warm).
Attention: Store, as supplied, at room temperature for up to 1 year. Store solutions at -20 °C for up to 3 months.
Formule:
C₁₉H₂₄N₂O Température de stockage: Température ambiante |
Numéro CAS:
6829-98-7 UN: 2811 ADR: 6.1,III |
|
Résultats des tests de spécification
Purity | ≥98% (TLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...