Imprimer
cis-5,8,11-Eicosatrienoic acid ≥97% (par HPLC), huile incolore
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): Mead acid
BMLFA0040001 BMLFA0040010
ENZOBMLFA0040001EA 165 EUR
ENZOBMLFA0040001 ENZOBMLFA0040010
cis-5,8,11-Eicosatrienoic acid ≥97% (par HPLC), huile incolore
cis-5,8,11-Eicosatrienoic acid
Immunomodulatory and anti-inflammatory effects.
Accumulates in tissues of essential fatty acid deficient animals. Is not converted to prostaglandins and LTB4 but is converted to cysteinyl leukotrienes. Immunomodulatory and anti-inflammatory effects.
Soluble in ethanol, DMSO, acetone, ethyl acetate or hexane.
Attention: Store, as supplied, at -20 °C for up to 1 year. Store solutions under an inert atmosphere at -20 °C for up to 3 months.
Formule: C₂₀H₃₄O₂
Poids moléculaire: 306.5 g/mol
Température de stockage: Congélateur
Numéro MDL: MFCD00065719
Numéro CAS: 20590-32-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf71b8a545872a75e11227024e22c7b795bcdf7b&vid=faf29be00a921d8777bccf4d5fec044155cd69fa&prid=web&clid=SAPDAM
Commander maintenant
Résultats des tests de spécification
Purity ≥97% (HPLC)