Imprimer
Bisindolylmaleimide I ≥98% (par CCM)
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): GF 109203X
BMLEI2460001 BMLEI2460005
ENZOBMLEI2460001EA 120 EUR
ENZOBMLEI2460001 ENZOBMLEI2460005
Bisindolylmaleimide I ≥98% (par CCM)
Bisindolylmaleimide I
Potent and selective protein kinase C inhibitor which has been used effectively in platelets, Swiss 3T3 fibroblasts and macrophages. Inhibitory profile: PKC, IC50=0.02µM (inhibits α, ßI, ßII and γ subtypes with similar potency); PKA, IC50=2.0µM; phosphorylase kinase, IC50=0.7µM; insulin, EGF and PDGF receptor tyrosine kinases are not inhibited at concentrations up to 50µM.
  • PKC inhibitor
Red solid.
Soluble in dimethyl formamide or DMSO (10mg/ml, warm to 40°C and vortex well). 
Formule: C₂₅H₂₄N₄O₂
Température de stockage: Congélateur
Numéro MDL: MFCD00236428
Numéro CAS: 133052-90-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36af12aac0eaeeddc87b91aca72c76847630a15c&vid=2fa7001a41f9c1aa5b3967f3c693e74ce6660450&prid=web&clid=SAPDAM