Imprimer
Bupropion hydrochloride ≥98% (par CCM)
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BMLNS1110050 BMLNS1110250
ENZOBMLNS1110050EA 84.7 EUR
ENZOBMLNS1110050 ENZOBMLNS1110250
Bupropion hydrochloride ≥98% (par CCM)
Bupropion hydrochloride
Inhibitor of dopamine and norepinephrine transporters
Inhibits dopamine and norepinephrine transporters (Ki=2.8 and 1.4µM respectively). Inhibits the serotonin transporter at higher concentration (Ki=45µM). In clinical use as an antidepressant and for smoking cessation. Also acts as a nicotinic receptor antagonist.
Formule: C₁₃H₁₉Cl₂NO
Numéro MDL: MFCD00055209
Numéro CAS: 34911-55-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7dd1435947ebe26d921e463614f804fbbb39b56&vid=2a6f147884768dfaa38abbb982c8c0b371bfb341&prid=web&clid=SAPDAM