Imprimer

Methanandamide ≥98% (par CCM)

Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BMLFA0210005 BMLFA0210025
ENZOBMLFA0210005EA 66 EUR
ENZOBMLFA0210005 ENZOBMLFA0210025
Methanandamide ≥98% (par CCM)
Methanandamide

Cannabinoid receptor ligand


Amidase resistant cannabinoid receptor (CB) agonist (CB1: Ki=20nM; CB2: Ki=815nM). The most potent of the series of methyl-anandamides. About 4-fold higher binding affinity for cannabinoid receptor CB1 than anandamide (Prod. No. BML-FA017) in the presence of PMSF. Does also bind to TRPV1 (Ki=4.67µM). Inhibits motor behavior resembling the effectts of THC rather than anadamide. 

Formule: C₂₃H₃₉NO₂
Poids moléculaire: 361.57 g/mol
Numéro MDL: MFCD00467914
Numéro CAS: 157182-49-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=237971e7a2d23e0f67f6d7b2b1010fc6c7c14e25&vid=83c677a9c4c0e852ae300a9d0cc536e62ae7ef1f&prid=web&clid=SAPDAM

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR