Imprimer

Picotamide ≥98%

Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Synonyme(s): 4-Methoxy-N1,N3-bis(pyridin-3-ylmethyl)isophthalamide

BMLRA1040010 BMLRA1040050
ENZOBMLRA1040010EA 110 EUR
ENZOBMLRA1040010 ENZOBMLRA1040050
Picotamide ≥98%
Picotamide

Thromboxane A2 antagonist and TXA2 synthase inhibitor


A dual activity thromboxane A2 antagonist and TXA2 synthase inhibitor. Clinically useful antiplatelet agent.

Formule: C₂₁H₂₀N₄O₃
Poids moléculaire: 376.416 g/mol
Numéro MDL: MFCD00866792
Numéro CAS: 32828-81-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73a2e6770ad287b966b369bd46ca62bac3a2ea4d&vid=b08864027af25bedd908d5ae3a69320724f0aff9&prid=web&clid=SAPDAM

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR