Imprimer

Chelerythrine chloride ≥98%

Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Synonyme(s): 1,2-Dimethoxy-12-methyl-[1,3]dioxolo[4',5':4,5]benzo[1,2-c]phenanthridin-12-ium chloride

BMLEI2250005 BMLEI2250025
ENZOBMLEI2250005EA 116 EUR
ENZOBMLEI2250005 ENZOBMLEI2250025
Chelerythrine chloride ≥98%
Chelerythrine chloride

PKC inhibitor


Reported to be a potent and selective inhibitor of protein kinase C (IC50=0.66µM). Although chelerythrine has numerous documented biological effects, studies have raised serious doubts about the effectiveness of chelerythrine as a PKC inhibitor.

Formule: C₂₁H₁₈ClNO₄
Poids moléculaire: 383.828 g/mol
Numéro MDL: MFCD00060717
Numéro CAS: 3895-92-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96b18c46455e9fd49dfd3afb0daab8a59726d741&vid=70cf371de9db9a542c51a74c7c28f7efcbe75bd4&prid=web&clid=SAPDAM

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR