Imprimer
Simvastatin ≥98% (par CCM)
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): (1S,3R,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-dimethylbutanoate
BMLG2440050 BMLG2440250
ENZOBMLG2440050EA 161 EUR
ENZOBMLG2440050 ENZOBMLG2440250
Simvastatin ≥98% (par CCM)
Simvastatin
HMG-CoA reductase inhibitor
HMG-CoA reductase inhibitor. Synthetic analog of Lovastatin (Prod. No. BML-G226). Inhibits the biosynthesis of isoprenoids and prevents small G protein activation. Displays lipid lowering and antihypertensive effects. Displays antiinflammatory, immunomodulatory effects as well as protective effects in experimental traumatic brain injury. Activates Rac1-dependent signalling pathway.
Formule: C25H38O5
Numéro MDL: MFCD00072007
Numéro CAS: 79902-63-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9623c52f40d73b09695fcf40beecb470b45d4337&vid=f9d3d6fedc4cb8adb520fb175b92ad84c306b6ef&prid=web&clid=SAPDAM