Imprimer

Vincristine sulphate ≥98%

Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BMLT1170005 BMLT1170025
ENZOBMLT1170005EA 161 EUR
ENZOBMLT1170005 ENZOBMLT1170025
Vincristine sulphate ≥98%
Vincristine sulphate

Microtubule inhibitor


Plant alkaloid that arrests the cell cycle in G2/M phase by blocking mitotic spindle formation. Depolymerizes microtubules and blocks binding of tubulin to microtubule proteins. Causes aberrant tubulin polymerization. Induces apoptosis. Triggers Raf-1 activation, phosphorylation of Bcl-2 family proteins and induction of p53 expression. Cancer chemotherapeutic agent. Posttranscriptionally downregulates HIF-1α expression.

Formule: C₄₆H₅₈N₄O₁₄S
Poids moléculaire: 923.04 g/mol
Numéro MDL: MFCD00084729
Numéro CAS: 2068-78-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3751449e4a5845753af6a8c3fd27b1a720b75321&vid=dc1d6c45f09c07ca2ffeb35558a455552f36b543&prid=web&clid=SAPDAM

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR