Imprimer
Capsaicine ≥99% (par CCM)
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): 8-Methyl-N-vanillyl-6-none-amid
BMLEI1250200 BMLEI1251000
ENZOBMLEI1250200EA 165 EUR
ENZOBMLEI1250200 ENZOBMLEI1251000
Capsaicine ≥99% (par CCM)
Capsaicine
TRPV1 agonist
Capsaicin is an agonist at the TRPV1 receptor, a ligand-gated non-selective cation channel. Exhibits neuroprotective effects in a model of transient global cerebral ischemia. Induces release of substance P from afferent nociceptive specific neurons. Constituent of cayenne pepper. Powerful excitant of peropheral sensory nerve endings, specifically unmyelinated afferent neurons (C-fibers). Reversibly inhibits aggregation of platelets.
Isolated from Capsicum  fruit.
Formule: C₁₈H₂₇NO₃
Poids moléculaire: 305.42 g/mol
Point de fusion: 66.1
Numéro MDL: MFCD00017259
Numéro CAS: 404-86-4
Indice Merck: 12,01811
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a732662048f287c9a63bcc5659c6d1a7e8e2d4c&vid=5f959a4140b10bc046c5a172d1195d1d307382be&prid=web&clid=SAPDAM