Imprimer
Quartz Glass Rods
Fournisseur: TECHNICAL QUARTZ SOLUTIONS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
75350 75351 75352 75353 75354 75355
TSLA75350EA 30.7 EUR
TSLA75350 TSLA75351 TSLA75352 TSLA75353 TSLA75354 TSLA75355
Quartz Glass Rods
Agitateurs Tiges d'agitation
Temperature resistant glass rods made from transparent quartz glass.
  • Chemical resistance
  • Made for tasks up to 1200 °C
  • High purity
TQS quartz glass rods stand for quality and versatile applications.