Imprimer
Height Adjustable Workstations
Fournisseur: Palbam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
WU1201 WU1001 WU1401
PALBWU1201EA 9480 EUR
PALBWU1201 PALBWU1001 PALBWU1401
Height Adjustable Workstations
Mobilier Tables
The cleanroom height adjustable workstation is designed with motorised lifters to allow the operator to quickly change the height of the table to ensure a comfortable, ergonomic work environment.
Table top and leveler base are manufactured from stainless steel 304 with electro-polished finish. The adjustable legs are manufactured from anodised aluminum. Height adjustability is controlled from easy to use front mounted control unit.
Table height can be adjusted from 760 to 1147 mm (30 to 45"). Custom dimensions are possible.
These tables are designed for 220 V.